Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-12
Entrez Gene ID
190306
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y55F3AM.2
Chromosome Location
Chr.IV: 1045983 - 1049506 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067628.5 NP_500029.2 1 - 2 103 91

View Full Product Details