Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srx-13
Entrez Gene ID
183437
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C44B12.4
Chromosome Location
Chr.IV: 1114697 - 1118666 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356907.1 NP_001343636.1 3 - 4 399 140

View Full Product Details