Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dsl-2
Entrez Gene ID
176938
Gene Name
Delta-like protein
Gene Aliases
CELE_W09G12.1
Chromosome Location
Chr.IV: 1202651 - 1204970 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067651.7 NP_500052.1 2 - 3 500 71

View Full Product Details