Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
npp-8
Entrez Gene ID
176966
Gene Name
protein-coding
Gene Aliases
CELE_Y41D4B.19
Chromosome Location
Chr.IV: 1567709 - 1595961 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356908.3 NP_001343633.1 9 - 9 2344 62

View Full Product Details