Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- nhr-92
- Entrez Gene ID
-
176971
- Gene Name
- Nuclear Hormone Receptor family
- Gene Aliases
-
CELE_Y41D4B.8
- Chromosome Location
-
Chr.IV: 1620094 - 1626584 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067710.7 |
NP_500111.3 |
9 - 10 |
2352 |
125 |
- Gene Symbol
- nhr-146
- Entrez Gene ID
-
6418682
- Gene Name
- Nuclear Hormone Receptor family
- Gene Aliases
-
CELE_Y41D4B.27
- Chromosome Location
-
Chr.IV: 1624063 - 1630756 on Build WBcel235
- Gene Symbol
- nhr-87
- Entrez Gene ID
-
176972
- Gene Name
- Nuclear Hormone Receptor family
- Gene Aliases
-
CELE_Y41D4B.7
- Chromosome Location
-
Chr.IV: 1624029 - 1630755 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383042.1 |
|
4 - 5 |
891 |
125 |
View Full Product Details