Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rps-28
Entrez Gene ID
176975
Gene Name
40S ribosomal protein S28
Gene Aliases
CELE_Y41D4B.5
Chromosome Location
Chr.IV: 1659659 - 1660042 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383043.1 1 - 2 151 89

View Full Product Details