Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ulp-3
Entrez Gene ID
3565434
Gene Name
ULP_PROTEASE domain-containing protein
Gene Aliases
CELE_Y48A5A.2
Chromosome Location
Chr.IV: 1963634 - 1964724 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028306.3 NP_001023477.1 1 - 2 296 70

View Full Product Details