Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cng-2
Entrez Gene ID
190732
Gene Name
protein-coding
Gene Aliases
CELE_Y76B12C.1
Chromosome Location
Chr.IV: 2037191 - 2043549 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307651.3 NP_001294580.1 1 - 2 321 71
NM_001356909.3 NP_001343690.1 11 - 12 2196 71

View Full Product Details