Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nsy-4
Entrez Gene ID
177019
Gene Name
Neuronal SYmmetry
Gene Aliases
CELE_Y38F2AL.1
Chromosome Location
Chr.IV: 2323265 - 2326775 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067788.8 NP_500189.4 2 - 3 322 67

View Full Product Details