Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppgn-1
Entrez Gene ID
177021
Gene Name
AAA domain-containing protein
Gene Aliases
CELE_Y38F2AR.7
Chromosome Location
Chr.IV: 2333832 - 2349040 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067790.7 NP_500191.3 10 - 11 2189 89
GenBank mRNA AB257343.1 - 9 - 10 2139 89

View Full Product Details