Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-82
Entrez Gene ID
177064
Gene Name
protein-coding
Gene Aliases
CELE_Y54G2A.8
Chromosome Location
Chr.IV: 2835664 - 2838723 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028336.4 NP_001023507.1 3 - 4 503 89
NM_001383059.1 3 - 4 487 89

View Full Product Details