Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- clec-85
- Entrez Gene ID
-
177068
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_Y54G2A.6
- Chromosome Location
-
Chr.IV: 2847206 - 2855456 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383060.1 |
|
8 - 9 |
2060 |
82 |
- Gene Symbol
- Y54G2A.40
- Entrez Gene ID
-
3896797
- Gene Name
- RING-type domain-containing protein
- Gene Aliases
-
CELE_Y54G2A.40
- Chromosome Location
-
Chr.IV: 2846476 - 2855643 on Build WBcel235
- Gene Symbol
- clec-84
- Entrez Gene ID
-
177067
- Gene Name
- C-type lectin domain-containing protein
- Gene Aliases
-
CELE_Y54G2A.33
- Chromosome Location
-
Chr.IV: 2842474 - 2855459 on Build WBcel235
View Full Product Details