Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eel-1
Entrez Gene ID
177084
Gene Name
protein-coding
Gene Aliases
CELE_Y67D8C.5
Chromosome Location
Chr.IV: 3032578 - 3050106 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307537.3 NP_001294466.1 5 - 6 3008 99
NM_001307538.3 NP_001294467.1 5 - 6 2936 99
NM_001307539.3 NP_001294468.1 5 - 6 3008 99
NM_001307540.3 NP_001294469.1 5 - 6 2867 99
NM_001307541.2 NP_001294470.1 5 - 6 3002 99
NM_001307542.3 NP_001294471.1 5 - 6 2867 99
NM_001307543.3 NP_001294472.1 5 - 6 3002 99
NM_067883.5 NP_500284.2 5 - 6 3000 99

View Full Product Details