The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001268315.3 | NP_001255244.1 | 4 - 5 | 471 | 107 |
NM_001383061.1 | 4 - 5 | 467 | 107 | ||
NM_001383062.1 | 4 - 5 | 469 | 107 | ||
NM_001383063.1 | 4 - 5 | 469 | 107 |