Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mca-3
Entrez Gene ID
177089
Gene Name
protein-coding
Gene Aliases
CELE_Y67D8C.10
Chromosome Location
Chr.IV: 3103152 - 3133149 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268315.3 NP_001255244.1 5 - 6 559 120
NM_001383061.1 5 - 6 555 120
NM_001383062.1 5 - 6 557 120
NM_001383063.1 5 - 6 557 120

View Full Product Details