Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dyf-3
Entrez Gene ID
177107
Gene Name
protein-coding
Gene Aliases
CELE_C04C3.5
Chromosome Location
Chr.IV: 3396581 - 3399231 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026125.3 NP_001021296.1 2 - 3 364 76
NM_001380144.1 NP_001367378.1 3 - 4 397 76
NM_001380145.1 NP_001367377.1 3 - 4 505 76

View Full Product Details