Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-23
Entrez Gene ID
186523
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_F58E2.9
Chromosome Location
Chr.IV: 3453608 - 3454788 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067949.2 NP_500350.2 2 - 3 592 73

View Full Product Details