Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-78
Entrez Gene ID
182469
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_C09G12.3
Chromosome Location
Chr.IV: 3491777 - 3493216 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067957.2 NP_500358.2 2 - 3 276 96

View Full Product Details