Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-79
Entrez Gene ID
182468
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_C09G12.2
Chromosome Location
Chr.IV: 3493610 - 3495179 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067958.4 NP_500359.1 1 - 2 166 152

View Full Product Details