Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-17
Entrez Gene ID
24105312
Gene Name
Vesicular acetylcholine transporter unc-17
Gene Aliases
CELE_unc-17
Chromosome Location
Chr.IV: 3618259 - 3624700 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383068.1 3 - 4 848 79
GenBank mRNA L19621.1 - 3 - 4 857 79

View Full Product Details