Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-20
Entrez Gene ID
186389
Gene Name
protein-coding
Gene Aliases
CELE_F56D6.5
Chromosome Location
Chr.IV: 3915596 - 3917083 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307706.2 NP_001294635.1 2 - 3 300 156

View Full Product Details