Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clec-76
Entrez Gene ID
189950
Gene Name
C-type lectin domain-containing protein
Gene Aliases
CELE_Y46C8AR.1
Chromosome Location
Chr.IV: 3965332 - 3969193 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068048.6 NP_500449.2 3 - 4 691 72

View Full Product Details