Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-4
Entrez Gene ID
184535
Gene Name
protein-coding
Gene Aliases
CELE_F15E6.7
Chromosome Location
Chr.IV: 4285403 - 4288432 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307353.2 NP_001294282.1 4 - 5 638 126
NM_068131.3 NP_500532.2 4 - 5 642 126

View Full Product Details