Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-15
Entrez Gene ID
187813
Gene Name
SET domain-containing protein
Gene Aliases
CELE_R11E3.4
Chromosome Location
Chr.IV: 4776364 - 4780229 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383087.1 7 - 8 1356 147

View Full Product Details