Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpf-5
Entrez Gene ID
177251
Gene Name
protein-coding
Gene Aliases
CELE_R11E3.8
Chromosome Location
Chr.IV: 4802702 - 4806586 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380210.1 NP_001368236.1 3 - 4 1100 72
NM_068246.7 NP_500647.2 4 - 5 1583 72

View Full Product Details