Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-95
Entrez Gene ID
186699
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_H06H21.2
Chromosome Location
Chr.IV: 4812075 - 4814029 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356919.2 NP_001343674.1 6 - 7 1158 74

View Full Product Details