Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gpa-18
Entrez Gene ID
189161
Gene Name
protein-coding
Gene Aliases
CELE_W03F8.9
Chromosome Location
Chr.IV: 5189007 - 5195631 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306789.3 NP_001293718.1 4 - 5 537 65
NM_001306790.3 NP_001293719.1 4 - 5 541 65

View Full Product Details