Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
thn-6
Entrez Gene ID
190415
Gene Name
THaumatiN family
Gene Aliases
CELE_Y59E9AL.1
Chromosome Location
Chr.IV: 5228919 - 5230138 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068346.2 NP_500747.1 1 - 2 131 100

View Full Product Details