Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nlp-47
Entrez Gene ID
185393
Gene Name
Neuropeptide-Like Protein
Gene Aliases
CELE_F36H12.1
Chromosome Location
Chr.IV: 5281768 - 5293689 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383093.1 2 - 3 203 83

View Full Product Details