Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rme-2
Entrez Gene ID
177329
Gene Name
protein-coding
Gene Aliases
CELE_T11F8.3
Chromosome Location
Chr.IV: 5469961 - 5473174 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068414.6 NP_500815.2 1 - 2 125 82

View Full Product Details