Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rme-2
Entrez Gene ID
177329
Gene Name
protein-coding
Gene Aliases
CELE_T11F8.3
Chromosome Location
Chr.IV: 5469961 - 5473174 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380231.1 NP_001368360.1 1 - 2 375 79
NM_068414.6 NP_500815.2 2 - 3 680 79

View Full Product Details