Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-17
Entrez Gene ID
182333
Gene Name
Serpentine Receptor, class V
Gene Aliases
CELE_C06E7.7
Chromosome Location
Chr.IV: 5867689 - 5870172 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068475.5 NP_500876.2 1 - 2 145 174

View Full Product Details