Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-7
Entrez Gene ID
177364
Gene Name
protein-coding
Gene Aliases
CELE_K02B2.4
Chromosome Location
Chr.IV: 5943981 - 5946638 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068493.8 NP_500894.1 2 - 3 729 126

View Full Product Details