Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-22
Entrez Gene ID
177369
Gene Name
protein-coding
Gene Aliases
CELE_C46G7.4
Chromosome Location
Chr.IV: 6019672 - 6033639 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306867.3 NP_001293796.1 7 - 8 1402 74
NM_001306868.2 NP_001293797.1 7 - 8 1402 74
NM_001306869.4 NP_001293798.1 7 - 8 1402 74
NM_001306870.4 NP_001293799.1 7 - 8 1402 74
NM_001306871.3 NP_001293800.1 6 - 7 1264 74
NM_001306872.3 NP_001293801.1 6 - 7 1264 74
NM_001306873.3 NP_001293802.1 6 - 7 1264 74
NM_001306874.3 NP_001293803.1 7 - 8 1519 74
NM_001306875.3 NP_001293804.1 8 - 9 1657 74
NM_001306876.3 NP_001293805.1 8 - 9 1657 74
NM_001306877.4 NP_001293806.1 8 - 9 1657 74
NM_001306878.3 NP_001293807.1 7 - 8 1519 74
NM_001306879.3 NP_001293808.1 7 - 8 1519 74
NM_001306880.3 NP_001293809.1 7 - 8 1519 74
NM_068505.4 NP_500906.2 6 - 7 1264 74
NM_068506.3 NP_500907.2 8 - 9 1657 74

View Full Product Details