Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpi-1
Entrez Gene ID
177372
Gene Name
Cystatin cpi-1
Gene Aliases
CELE_K08B4.6
Chromosome Location
Chr.IV: 6092603 - 6093680 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068514.6 NP_500915.1 2 - 3 228 86

View Full Product Details