Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-30
Entrez Gene ID
188461
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T13A10.7
Chromosome Location
Chr.IV: 6267457 - 6269446 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372839.1 NP_001360069.1 5 - 6 888 168

View Full Product Details