Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
htp-2
Entrez Gene ID
177405
Gene Name
HORMA domain-containing protein
Gene Aliases
CELE_Y73B6BL.2
Chromosome Location
Chr.IV: 6419500 - 6420898 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068580.7 NP_500981.3 3 - 4 975 68

View Full Product Details