Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lgc-10
Entrez Gene ID
177409
Gene Name
protein-coding
Gene Aliases
CELE_Y73B6BL.26
Chromosome Location
Chr.IV: 6446542 - 6457572 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306849.3 NP_001293778.1 7 - 8 828 96
NM_068584.6 NP_500985.4 6 - 7 733 96

View Full Product Details