Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
try-3
Entrez Gene ID
183420
Gene Name
protein-coding
Gene Aliases
CELE_C43G2.5
Chromosome Location
Chr.IV: 6557155 - 6560955 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380280.2 NP_001367393.1 1 - 2 210 108

View Full Product Details