Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- best-9
- Entrez Gene ID
-
183419
- Gene Name
- Bestrophin homolog 9
- Gene Aliases
-
CELE_C43G2.4
- Chromosome Location
-
Chr.IV: 6557328 - 6565270 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383107.1 |
|
4 - 5 |
417 |
75 |
- Gene Symbol
- try-3
- Entrez Gene ID
-
183420
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C43G2.5
- Chromosome Location
-
Chr.IV: 6557155 - 6560955 on Build WBcel235
View Full Product Details