Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
klp-12
Entrez Gene ID
178057
Gene Name
protein-coding
Gene Aliases
CELE_T01G1.1
Chromosome Location
Chr.IV: 11341325 - 11359869 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028178.4 NP_001023349.1 16 - 17 3590 130
NM_001129324.3 NP_001122796.1 16 - 17 3590 130

View Full Product Details