Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cbd-1
Entrez Gene ID
178061
Gene Name
Chitin-binding domain protein cbd-1
Gene Aliases
CELE_H02I12.1
Chromosome Location
Chr.IV: 11372991 - 11377761 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069744.7 NP_502145.2 1 - 2 44 63

View Full Product Details