Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-25
Entrez Gene ID
192076
Gene Name
protein-coding
Gene Aliases
CELE_M04B2.5
Chromosome Location
Chr.IV: 11504716 - 11509727 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276863.2 NP_001263792.1 5 - 6 900 107
NM_069769.5 NP_502170.4 14 - 15 1914 107
NR_101961.1 14 - 15 1862 107

View Full Product Details