Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- srgp-1
- Entrez Gene ID
-
178079
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F12F6.5
- Chromosome Location
-
Chr.IV: 11570500 - 11578699 on Build WBcel235
- Gene Symbol
- F12F6.1
- Entrez Gene ID
-
178081
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_F12F6.1
- Chromosome Location
-
Chr.IV: 11570500 - 11588322 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069781.8 |
NP_502182.2 |
4 - 5 |
2988 |
101 |
View Full Product Details