Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prx-2
Entrez Gene ID
178094
Gene Name
RING-type domain-containing protein
Gene Aliases
CELE_ZK809.7
Chromosome Location
Chr.IV: 11644129 - 11646322 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069800.5 NP_502201.2 1 - 2 272 69

View Full Product Details