Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-8
Entrez Gene ID
178101
Gene Name
Innexin-8
Gene Aliases
CELE_ZK792.2
Chromosome Location
Chr.IV: 11673843 - 11675636 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069808.6 NP_502209.1 3 - 4 1111 73

View Full Product Details