Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-9
Entrez Gene ID
191694
Gene Name
Innexin
Gene Aliases
CELE_ZK792.3
Chromosome Location
Chr.IV: 11676354 - 11678497 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069809.3 NP_502210.2 1 - 2 162 70

View Full Product Details