Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lpr-2
Entrez Gene ID
178111
Gene Name
LiPocalin-Related protein
Gene Aliases
CELE_T12A7.5
Chromosome Location
Chr.IV: 11779216 - 11786975 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069830.5 NP_502231.1 15 - 16 2262 92

View Full Product Details