Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhx-9
Entrez Gene ID
178126
Gene Name
protein-coding
Gene Aliases
CELE_ZK822.3
Chromosome Location
Chr.IV: 11930872 - 11935719 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028456.4 NP_001023627.1 3 - 4 662 139
NM_001028457.5 NP_001023628.1 3 - 4 340 139
NM_001383212.1 3 - 4 472 139
GenBank mRNA AF497834.1 - 3 - 4 362 139
AF497835.1 - 3 - 4 614 139

View Full Product Details