Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbx-36
Entrez Gene ID
191435
Gene Name
Putative T-box protein 36
Gene Aliases
CELE_ZK829.5
Chromosome Location
Chr.IV: 11950570 - 11952208 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069865.4 NP_502266.1 1 - 2 186 146

View Full Product Details